Vernier® Surface Temperature Sensor
Supplier: Vernier Software & Technology
STS-BTA
470017-598EA 35.69 USD
470017-598
Vernier® Surface Temperature Sensor
Temperature Probes
Vernier Surface Temperature Sensor provides a rapid response time.
  • Quick Readings
  • Exposed Thermistor Provided
  • For Use in Situations Requiring Flexibility or Low Thermal Mass
  • Maximum Temperature the Sensor Tolerates without Damage: 150°C
  • Applicable to Experiments Including Specific Heat, Skin Temperature, and More
This sensor features an exposed thermistor that helps provide accurate readings quickly and efficiently. It can assist in a wide variety of experimentation including specific heat, friction and energy, skin temperature, and more.
The sensor is for educational use in air and water only.
Resolution: 0.08°C (–25 to 0°C), 0.03°C (0 to 40°C), 0.1°C (40 to 100°C), 0.25°C (100 to 125°C)
Response time: For 90% change in reading: 50 seconds (in still air), 20 seconds (in moving air)