Vernier® Soil Moisture Sensor
Supplier: Vernier Software & Technology
SMS-BTA
470017-560EA 155.39 USD
470017-560
Vernier® Soil Moisture Sensor
Electrodes Other Electrodes
The soil moisture sensor is used to measure the volumetric water content of soil. This makes it ideal for performing experiments in courses such as soil science, agricultural science, environmental science, horticulture, botany, and biology.
  • Dimensions: 8.9×1.8×0.7 cm (active sensor length 5 cm)
  • Power: 3 mA @ 5VDC
Simply insert it into the soil to determine the volumetric water content in percent. Resolution: 0.1%.