Ward's® Single Probes Voltage Sensor
Supplier: Wards
WSP-101-500
470017-546EA 74.65 USD
470017-546
Ward's® Single Probes Voltage Sensor
Electrodes Other Electrodes
Introduce and explore electricity concepts and safely measure electrical voltage with the Ward's Single Probes Voltage Sensor.
  • Explore concepts of electricity
  • Two banana plugs
  • Free activity guide
This sensor easily and accurately measures the voltage of electronic circuits using two banana plugs, included with the sensor. Sensor includes free activity guide with activities designed for use with each probe, available digitally online after purchase. Range: -20 to 20V; Resolution: 0.01V. Max sample rate 10,000 per second. Requires Ward's Single Probes Base Unit, available separately.