SPECTRONIC™ 200 Spectrophotometer
Supplier: Thermo Scientific
840-282500
470017-544EA 2462.09 USD
470017-544
SPECTRONIC™ 200 Spectrophotometer
Spectrophotometers and Photometers UV/Visible Spectrophotometers
No Computer or Serial Cable Required; Just Plug It in and You’re Ready to Measure.
  • With no internal parts, there’s nothing to wear out
  • Compact footprint saves space in your lab
  • Large, bright, color LCD adjusts to the perfect angle for any technician to view
  • Screen-print feature gives a permanent record of your data
  • Quantitative analysis with either a factor or up to four standards offers confidence
Thermo Scientific™ SPECTRONIC 200™ is the new standard for rugged, simple-to-use visible spectrophotometers for teaching and routine applications. The new SPECTRONIC 200 spectrophotometer couples the simplicity and reliability of the Thermo Scientific™ SPECTRONIC 20 and the Thermo Scientific ™ GENESYSTM 20 instruments with 21st century technology and a bold new design that promises years of trouble-free performance. On-board software, controlled from an intuitive navigation pad using a full color screen, delivers capabilities beyond expectations.