Milwaukee Economy Portable Digital Turbidity Meter
Supplier: MILWAUKEE INSTRUMENTS, INC.
MI415
470017-306EA 901 USD
470017-306
Milwaukee Economy Portable Digital Turbidity Meter
Turbidity Meters
Determine How Many Particulates are in the Stream
  • No Manual Calibration Necessary
Accurate turbidity measurement is achieved with this meter by analyzing the amount of light refracted from suspended particles such as clay, silt and organic material. The meter features two operating ranges: 0.00 to 50.00 FNU and 50 to 1000 FNU to accomodate even the most turbid of conditions. Easy to use, this unit is factory programmed, so no manual calibration is necessary. Supplied with two cuvets, reagents, hard carrying case, wiping tissue, 9V battery, and instructions. Range: 0.00 to 50.00 FNU; 50 to 1000 FNU. Resolution: 0.01 FNU; 1 FNU. Accuracy: ±0.5 FNU or ±5% of reading, whichever is greater. Environment: 0–50°C / 32–122°F; max RH 100%. Size: 192 x 104 x 52 mm. Weight: 380 g.
Turbidity Range: 0.00 - 50.00 FNU; 50 - 1000FNU. Resolution: 0.01 FNU; 1 FNU. Accuracy: ±0.5 FNU; ±5% of reading.