Essential Physics Demo: Wood and Metal Heat Conducting Cylinder
Supplier: Wards
PH0405/PK5
470017-140KT 29.9 USD
470017-140
Essential Physics Demo: Wood and Metal Heat Conducting Cylinder
Educational Materials Physics Educational Materials Heat/Thermodynamics Learning Activities
Demonstrate the Difference Between the Specific Heat of Objects.
  • Affordable, Easy, Fun
  • Present these demonstrations in front of your class to excite your students about learning core physics concepts
  • Set up and execution are simple, so you can spend more time engaging your students
  • Ideal for introducing a unit or using as reinforcement after a lesson, these demos are available for all fundamental physical science topics
This 19cm cylinder, which is constructed of wood on one end and hollow metal on the other, enables you to show how metal has a lower specific heat than wood in several ways. One way is to simply let your students feel both ends of the cylinder at room temperature, enabling them to see that the metal end is cooler than the wooden end. Another easy demonstration is to wrap paper on both ends and hold the cylinder over a flame for five seconds. The metal end will be relatively unmarked while the wooden end will show scorch marks. Included are five cylinders and instructions.