Quantitative Spectroscope
Supplier: Wards
PH0595
470017-108EA 87.4 USD
470017-108
Quantitative Spectroscope
Educational Materials Physics Educational Materials Optics Learning Activities
Excellent for student use.
  • Incredibly durable and attractive metal housing
  • Includes detachable cuvette holder, with cuvettes
  • Simple to use
This spectroscope features a holographic replica grating of 600 lines per mm. which enables for detailed analysis through the high quality resolution. Students can calibrate the bright bands of the dominant wavelengths in nanometers using the included internal scale, and also insert and analyze liquid samples with the three included cuvettes.