Laboratory Safety Mat
Supplier: Fischer Technical
MATRED
470016-916EA 56 USD
470016-916
Laboratory Safety Mat
Surface Protectors Bench Protectors
Stabilize glassware on orbital or rocking shakers and eliminate slippery conditions on benchtop surfaces with this versatile non-slip safety mat.
Its ribbed, textured surface steadily holds beakers, cylinders, and other glassware in place, even at an angle. A valuable addition to any laboratory, the silicone rubber mat can be autoclaved, cleaned with disinfectants such as bleach, easily cut to various sizes, and will withstand temperatures up to 260°C.
Size: 9 1/2" square x 1/8" thick.