Wards® QA Spectroscope
Supplier: VWR International
PH0597-N11
470016-874CS 100 USD
470016-874
Wards® QA Spectroscope
Educational Materials Physics Educational Materials Optics Learning Activities
This semi-quantitative spectroscope is a simple and economical alternative to expensive equipment.
  • Perfect for Chemistry and Physics classrooms
  • Inexpensive and simple to use
  • Instructions included
The Ward's® QA spectroscope has been designed to produce a bright spectrum of visible light decomposed by a diffraction grating, with a clear, easy-to-view translucent scale. Students can analyze spectra from flames and gas emission tubes, measuring wavelengths from 4000 to 7000 Å with an accuracy of +/-50 Å. The spectroscope houses a 37 mm scale, and includes instructions with suggestions for use.