Giant Color Coded Periodic Table
Supplier: BRITTEN BANNER INC.
4751300
470016-786EA 119.65 USD
470016-786 470039-668 470104-090
Giant Color Coded Periodic Table
Charts
Large enough to be seen from ANY corner of the classroom.
  • Also available in 8.5 x 11" student pads and transparency
  • Uses IUPAC and older CAS numbering
This oversized periodic table clearly displays all elements, including synthetic and radioactive elements. Color coding makes it easy for your students to distinguish between metals, nonmetals, metalloids and groups. Table uses both the new IUPAC group numbering system (1–18) and the older CAS numbering system (IA, IIA, IIIB, VIB...). Details given for each element include name, symbol, atomic number, atomic weight, and a state of matter at 20°C. Also included is a table of naturally occurring isotopes and a color coded table of common uses.