Nylon Semimicro Test Tube Brush
Supplier: Mill-Rose
73200B
470016-166EA 3 USD
470016-166 470120-526
Nylon Semimicro Test Tube Brush
Brushes
For test tubes 5 to 11 mm in diameter. Total length: 14.6 cm; length of fiber section: 7 cm; diameter: 1.3 cm.