Handmade Rubber Tubing
Supplier: Prestige Rubber
00516011610R
470016-106PK 19.7 USD
470016-106
Handmade Rubber Tubing
Tubing Transfer Tubing
Seamless tubing made of pure gum rubber except for the pigment. Will retain its high elasticity for years; recommended for laboratory use.
  • Specific gravity: 0.97