Interference Model
Supplier: LD DIDACTIC GMBH
415 04
470016-066EA 92.7 USD
470016-066
Interference Model
Educational Materials Physics Educational Materials Optics Learning Activities
The concentric circles on each of these two transparent acetate plates help make the concept of wave interference perfectly clear.
  • One plate is 9 cm x 9 cm; the other is 9 cm x 12 cm
When the plates are laid on top of one another and moved, the circles, representing wavefronts, create visually dramatic patterns of interference. The plates can be used on an overhead projector.