Pulley Demonstration Set
Supplier: Wards
PH0305
470016-048EA 387.14 USD
470016-048
Pulley Demonstration Set
Educational Materials Physics Educational Materials Mechanics Learning Activities
A simpler version of the Advanced Pulley Demonstration Set, this model includes eight single pulleys and two double sheave pulleys in addition to EISCO ball bearing frictionless pulley.
  • Economical
  • Ball-Bearing, Frictionless Pulley
  • Wooden Base
  • Suitable for Introductory Pulley Demonstrations
A wooden base supports one vertical rod with attachments to a horizontal rod at top.
This set has everything needed to conduct a series of introductory pulley demonstrations.
Ordering information: This kit includes hooked masses, collars with hooks, clamps, and cord.