Deluxe Loop the Loop
Supplier: Shiv Dial Sud & Sons
P10470A
470016-032EA 46 USD
470016-032
Deluxe Loop the Loop
Educational Materials Physics Educational Materials Mechanics Learning Activities
Instantly transform potential energy into kinetic energy.
  • Size: 110 cm L
A riveting demonstration of conservation of energy, conservation of momentum, and projectile motion, the Loop the Loop looks like the track of a rollercoaster. The curved metal track allows you to drop a steel ball from the top and watch the results. Depending on the height at which it is dropped the ball will roll completely around the interior of the track or will drop to the ground. A circular scale is attached to the device for measuring the angle at which the ball drops. Includes two steel balls and instructions.