AC/DC Universal Power Supply
Supplier: Wards
ESRUV1339
470016-026EA 480.3 USD
470016-026
AC/DC Universal Power Supply
Power Supplies
AC/DC power supply with overload protection.
  • Overload protection indicator
  • Glass fuses at the input level
  • One spare fuse at the mains input socket
This power supply provides seven DC voltages and one AC voltage. The 5V, 8V, and -8V are the regulated voltages provided, and all others are unregulated filtered DC voltages. All AC and DC voltages can be used simultaneously if desired.
Certifications: CSA approved.