C Clamps
Supplier: United Scientific Supplies
CLMPC3 #044300 CLMPC4
470005-368EA 15.9 USD
470005-368 470015-954 470104-094
C Clamps
Clamps General Purpose Clamps
Securely hold two flat object together.
  • Cast iron frame
  • Screw thread mechanism
These cast iron C clamps help to securely holes surfaces together. The maximum opening of the jaw needs to be taken into account when selecting the correct size. The horizontal torque bar allows you to apply a lot of force to tighten or release.