Hair Dryer
Supplier: OSI Chicago
3703
470015-950EA 73.6 USD
470015-950
Hair Dryer
Heaters Hot Air Dryers
An ergonomic handle and two heat/speed settings.
  • 1875 watts
  • Hang ring
  • 5-ft. power cord
For drying glassware, electrostatic equipment, chromatograms and precipitates, and for many other laboratory uses.