Bone, Ground Preparation Slide
Supplier: Avantor
470015-864EA 14.49 USD
470015-864
Bone, Ground Preparation Slide
Slides Prepared Slides
Examine bone tissue microscopically
  • Identifying features clearly distinguishable
  • Rigorous quality control standards
Human. Thin section showing lacunae distinct with canaliculi.