Tryptic Soy Broth
Supplier: Avantor
470015-844EA 27.95 USD
470015-844
Tryptic Soy Broth
Media for Microbiology Dehydrated Media for Microbiology
Soybean-casein digest medium. Supports the growth of a variety of microorganisms.
Premeasured Packs Save Time in the Lab
Simply add the contents of one Dehydrated Media pack to one liter of water, bring to a boil, and sterilize, to make enough media for 40 to 50 plates or 16edited0 tubes.