Hygro–Thermometer
Supplier: Sper Scientific
800016
470015-832EA 79.46 USD
470015-832
Hygro–Thermometer
Hygrometers Digital Thermo-Hygrometers
Battery operation makes the unit completely portable.
  • Records min/max temperature and humidity
  • Can be mounted to the adhesive wall hanger
  • Aligned with GLOBE protocols
Because it’s battery–powered, you can use the thermometer/hygrometer for virtually any application requiring careful monitoring of temperature and humidity. The large digital display also recalls minimum/maximum temperature and humidity readings over any time period.
Temperature range: 32 - 122 °F (0 - 50 °C) with accuracy of ±2 °F or 1 °C and a 1° resolution for both °F and °C
Relative humidity range: 20 - 99% with an accuracy of ±3% and a resolution of 1%. One AAA battery and an adhesive wall hanger are included.