Somso® Ear Model
Supplier: MARCUS SOMMER
DS 5
470015-738EA 982.2 USD
470015-738
Somso® Ear Model
Models Human Biology Models
Dissectible To The Most Detailed Individual Structures
  • 3X Size Is Great For Lab group Or Station Usage
  • Important Structures Are Easily Viewable
  • 5 Year Manufacturers Warranty
Enlarged to 3X life-sized, this model can be separated into six pieces. Removable pieces include the petrous portion of the temporal bone, a section of the auditory canal, the labyrinth, the tympanic membrane, the malleus, and the incus.