Somso® Earthworm Model
Supplier: MARCUS SOMMER
ZOS 108
470015-716EA 1123.3 USD
470015-716
Somso® Earthworm Model
Models Zoology Models
Most Detailed, Intricate Rendering Of Earthworm Anatomy Available
  • Highly Detailed Dissection
  • Removes From Stand For Easy Examination
  • 5 Year Manufacturers Warranty
Enlarged 25X and colorfully illustrated, the comprehensive model can be taken apart for an even closer view of earthworm structure. It features a sectioned and dissected anterior to the clitellum to clearly show the septa, coelom, nephridia, aortic arches or “hearts”, and even the setae. The intestine is removable for a better view of the stomach, and the reproductive organs are removable to show the sperm ducts.