Somso® Spinal Cord with Nerve Branches Model
Supplier: MARCUS SOMMER
BS 32/37
470015-608EA 452.88 USD
470015-608
Somso® Spinal Cord with Nerve Branches Model
Models Human Biology Models
Set Of Two Models Shows Origin Of Spinal Nerves
  • 2 Models Show Varying Aspects Of The Spinal Cord
  • Key Stored In Base For Easy Access
  • 5 Year Manufacturers Warranty
Provide students with a unique look at the central nervous system with this set of two models. The spinal cord section, enlarged 10X, details the white and gray matter, sulci, ganglion, and fissures. The model of the spinal cord with nerve branches, enlarged 5X, displays the ventral and dorsal roots of the spinal nerves. The two models are mounted on a common base, and a transparent case is included for protection.