Ward's® Crystal Form Collection
Supplier: Avantor
470015-404EA 40.69 USD
470015-404
Ward's® Crystal Form Collection
Specimens Mineral Specimens Crystals
Basic Set of Natural Crystals
  • Hand Selected by WARD's Geologists
  • Teach Students the Basic Crystal Forms
The six main crystal systems are represented in this beginning geology student’s introduction to crystal form. The set is housed in a compartmented collection box, and an identification list is included.
Contents:6 numbered samples, 1" x 1"
Isometric Pyrite
Hexagonal Quartz
Tetragonal Zircon
Orthorhombic Danburite
Monoclinic Orthoclase
Triclinic Microcline