TriMan Overhead Compass/Ruler
Supplier: SI Manufacturing
SIG56079
470014-902EA 5 USD
470014-902
TriMan Overhead Compass/Ruler
Rulers
It is versatile and easy to use, this precision tool allows you to draw circles, straight lines, and complex geometric shapes with accuracy and artistry.
Construct, copy, and read angles; measure and convert inches and centimeters; and much more. To use this device, simply slide the cursor to the desired position and lock; placing a finger on the transparent swivel disk will anchor the ruler as you draw with the pencil point in the cursor hole.
WARNING: CHOKING HAZARD- Small Parts. Not for Children Under 3 Years.