Ward's® Single Pressure Probe
Supplier: Wards
WSP-106-500
470014-830EA 132.4 USD
470014-830
Ward's® Single Pressure Probe
Pressure Sensors
Explore properties of gas and easily incorporate hands-on digital data collection with the WARD'S Single Pressure Probe.
  • Explore properties of gas
  • Durable sensor
  • Connects to many containers
This durable sensor connects easily to a variety of containers and apparatuses, making it easy to use with any lesson. Range: 0 to 7 atm, 0 to 100 psi, 0 to 700 kPa, 0 to 7 bar. Resolution: 0.01 atm, 0.1 psi, 0.1 kPa, 0.01 bar.
Ordering information: Probe requires WARD'S Single Pressure Base Unit, available separately.