Ward's® Single Probes Light Sensor
Supplier: Wards
WSP-104-500
470014-820EA 85.75 USD
470014-820
Ward's® Single Probes Light Sensor
Temperature Probes
Measure the intensity of light in the darkest of classrooms to brightest school yards with the Ward's Single Probes Light Sensor.
  • Measure the intensity of light
  • Collect and record data
  • Free activity guide
It's easier than ever for students to collect and record light data for experimentation and analysis. Sensor includes free activity guide with activities designed for use with each probe, available digitally online after purchase. Resolution: 0-1000 lx 1 lx, 0-6000 lx 1 lx, and 0-150,000 lx 22 lx. Sample rate, 10,000. Requires Ward's Single Probes Base Unit, available separately.