Ward's® Single Probes Current Sensor
Supplier: Wards
WSP-102-500
470014-778EA 74.95 USD
470014-778
Ward's® Single Probes Current Sensor
Electrodes Other Electrodes
Introduce and explore electricity concepts and safely measure electrical current with the Ward's Single Probes Current Sensor.
  • Explore concepts of electricity
  • Two banana plugs
  • Free activity guide
This sensor easily and accurately measures the current of electronic circuits using two banana plugs, included with the sensor. Sensor includes free activity guide with activities designed for use with each probe, available digitally online after purchase. Range: -2.5 to 2.5 amps; Resolution: 1 mA. Sample rate, 10,000 per second. Requires Ward's Single Probes Base Unit, available separately.