Soil/Sediment Sifter
Supplier: Science First
621-7120
470014-728EA 57.89 USD
470014-728
Soil/Sediment Sifter
Sieves Soil Sampling Sieves
Ideal for soil and general particle sifting, this sifter’s 2mm aperture (2000 micron) corresponds to sieve size 10. The high-quality, durable wood frame and steel mesh will last through years of student use. Size: 24" x 16" x 3".