Human Genetics Taste Paper Set
Supplier: Avantor
470014-676KT 22.95 USD
470014-676
Human Genetics Taste Paper Set
Educational Materials Biology Educational Materials Genetics Learning Activities
By using three different chemicals, students and relations can get their genetically determined taste responses tested.
The set includes pedigree orms, WL4539P and four 100-paper vials, one each of: PTC, sodium benzoate, thiourea and a control paper. The four different test papers are also sold separately.