White Light Illuminator
Supplier: PROMARK INTERNATIONAL, INC.
750428
470014-592EA 189.5 USD
470014-592
White Light Illuminator
Transilluminators White Light Transilluminators
Provides the Perfect Amount of Light for Photographing Gels
With its large surface area and even distribution of white light, the large light table is ideal for viewing or photographing DNA or PAGE electrophoresis gels. 110/240 VAC with a 10' cord. Viewing surface size: 9 1/2"L x 6"W. Overall size: 9"L x 12 1/2"W x 2 1/4"H.