Function Generator, 100 kHz
Supplier: VWR International
PH1296-N9
470014-358EA 373.6 USD
470014-358
Function Generator, 100 kHz
Batteries
Versatile signal generator.
  • Generator operates on 120V/240V AC using voltage selector switch
  • 0–10V peak to peak
Output waves are measured in sine, square, and triangular form. Amplitude is 0–10V peak to peak in three steps with coarse and fine adjustment. AC input is protected by a fuse of 0.5A, and output is short circuit protected.