DC Battery Eliminator, 1 Amp
Supplier: Wards
PH0971A-V1
470014-352EA 142.42 USD
470014-352
DC Battery Eliminator, 1 Amp
Power Supplies
This battery eliminator features overload protection.
  • The Unit Runs Any Low-Power Apparatus
  • Output Via Two Insulated Binding Posts
  • Short Circuit Protection
  • Overload Protection
  • Line and Load Regulation
The design includes overload and short circuit protection as well as line and load regulation.
Switching technology results in a compact, lightweight unit. The unit runs any low-power apparatus.