Fundamentals of Physics Optics Bench
Supplier: Wards
FN-OPTKIT
470014-334EA 166.4 USD
470014-334
Fundamentals of Physics Optics Bench
Educational Materials Physics Educational Materials Physics Fundamentals Learning Activities
Convert the fun stand into an optical bench.
  • Large colored rods
  • Includes teacher manual
Offer your students a hands-on experience with geometric optics, demonstrating polarization, color mixing, and more. The unique design of this device uses supports that attach to the Fun Stand as a horizontal bar and create a student-friendly optical bench for many experiments. Instructions for all activities are also supplied.
Accessories information:
Requires, the Fun Stand, available separately. Requires a 12 V power supply, not included.
Delivery information:
Kit Includes: (4) color filters; (2) polarizing lenses; support brackets; light source; support stand; and (2) each of concave and convex lenses, and mirrors; all securely organized in a sturdy storage container.