Ward’s® Organic/Inorganic Molecular Model Student Set, 95 Pieces
Supplier: VWR International
470014-294
470014-294EA 44.58 USD
470014-294
Ward’s® Organic/Inorganic Molecular Model Student Set, 95 Pieces
Models Molecular Biology Models
Useful for individual or group study of molecules, these model sets offer an interactive learning opportunity.
  • Models Useful for Individual or Group Study
  • Interactive Learning Opportunity
  • Cost-Effective
  • Convenient Plastic Case Included
  • Instructions Included
These model sets offer an interactive learning opportunity. Students construct molecules from color-coded plastic atoms and learn about the structure and function of each. The holes in each atom are set at angles that accept the included, flexible connectors of various lengths that represent the different bonds. Medium, gray links represent single covalent bonds, long and gray links represent double and triple covalent bonds, and short and white links can be used to construct compact models. With this comprehensive large set, students learn about alkanes, alcohols, aldehydes, ketones, and other organic structures.
Students can now construct both inorganic and organic chemistry structures with one comprehensive set. In addition to common organic molecules, this set allows you to build metal oxides, hydroxides, non-metal oxides, hydrides, and halides. The complete set with instructions comes in a convenient plastic storage case.
Ordering information: 6 carbon black and 4-holed 22 mm atoms, 14 hydrogen white and 1-holed 22 mm atoms, 2 nitrogen blue and 4-holed 22 mm atoms, 6 oxygen red and 2-holed 20 mm atoms, 1 sulfur yellow and 2-holed 22 mm atoms, 1 sulfur yellow and 4-holed 22 mm atoms, 1 sulfur yellow and 6-holed 22 mm atoms, 1 phosphorus purple and 5-holed 22 mm atoms, 6 halogen green and 1-holed 20 mm atoms, 3 metal grey and 1-holed 15 mm atoms, 3 metal grey and 2-holed 22 mm atoms, 2 metal grey and 3-holed 22 mm atoms, 1 metal grey and 4-holed 22 mm atoms, 1 brown and 4-holed 22 mm atoms, 1 brown and 5-holed 22 mm atoms, 1 brown and 6-holed 22 mm atoms, 25 medium links, and 10 long and flexible links.