BlotBoy™ 3D Platform Rocker
Supplier: Ward's
SWBR1000-S
470013-890EA 124.4 USD
470013-890
BlotBoy™ 3D Platform Rocker
Shakers and Mixers Rotating/Rocking Shakers
Gentle Gyratory Mixing Action Ideal for Blotting and Gel Work
Safe for use in cold rooms and incubators from 4–65°, this platform rocker provides the perfect 3D motion for all blot related applications, gel staining, and destaining. Its speed and ±8° tilt angle have been optimally set at 12rpm for gentle yet thorough mixing in gel trays, boxes, and other flat vessels. Includes autoclavable flat mat, 12" x 12" platform, and two year warranty. Rocker can support up to 4.4 lbs. 115 VAC. CSA compliant. Size: 12"W x 12"D x 8 1/4"H; weight: 12kg.
A Stacking Platform is also available separately.More Product Information