Wind Vane Demonstrator, SCIENCE FIRST INC SE
Supplier: Science First
652-1020
470013-618EA 32.4 USD
470013-618
Wind Vane Demonstrator, SCIENCE FIRST INC SE
Anemometers
Simple to operate and easy to use, this economical wind vane clearly provides current wind direction. Its heavy cast base and sturdy post support a sensitive cone bearing assembly that responds to even the slightest wind. Place the “N” portion of the wind vane in the northern position and read off the direction shown by the pointer.