Pocket Soil Moisture Meter
Supplier: General Tools Manufacturing
GLMM200
470013-574EA 27.8 USD
470013-574
Pocket Soil Moisture Meter
Analyzers Moisture Analyzers
Sized conveniently to carry in the field.
  • Aligned with GLOBE protocols
Test the conditions of local soil with this heavy-duty probe. Instant readings are given for moisture content, and no batteries are required for operation. A plant water chart is also included. Size: 10"W x 21/4"H x 1"D.