Spinplus® Magnetic Stirring Bars
Supplier: Bel-Art
F37144-0100_SE
470013-464EA 32.3 USD
470013-464
Spinplus® Magnetic Stirring Bars
Stirrers Magnetic Stirring Bars
Spinplus® magnetic stirring bars add speed and efficiency to mixing operations.
  • The “+” shape creates a deep vortex and provides stable, quiet operation
  • Alnico magnet encapsulated in FDA Grade, USP Class VI Teflon® PTFE low friction coating; durable and inert
  • Manufactured in a registered ISO 9001:2008 facility, as verified by SGS certification
  • Autoclave safe