Mini ProBath Waterbath

Supplier: Revolutionary Science
RS-PB-50
470013-450EA 306.55 USD
470013-450
Mini ProBath Waterbath
Baths Water Baths, Unstirred
A fraction of the cost of other water baths, the Mini ProBath Water bath is as versatile as larger units.

  • Compact Model, 0.5 L
  • Diameter: 8"
  • Height: 4½"
  • Ideal for Warming Agar

Ideal for warming agar, the water bath can also be used to create a steam bath or keep media warm.

Its temperature range is ambient–100°C.

Ordering information: This water bath includes a heating-indicator light, temperature regulator control knob, and transparent polymer cover.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR