Ohaus® YA Series Pocket Scales
Supplier: OHAUS CORP.
80251910
470013-376EA 159.8 USD
470013-376
Ohaus® YA Series Pocket Scales
Balances and Scales
YA series pocket scales offer portable precision weighing in a compact case.
  • Fast stabilization time
  • Backlit LCD display
  • Power-saving auto shut-off feature
  • One-year warranty
Put convenient, compact, and portable weighing capability right in the palm of your hand. These pocket sized scales deliver great performance in a stylish, yet functional design. With simple operation, the units include a stainless steel weighing pan and unique hinged cover to protect the scale. The rugged plastic scales weigh in g, oz, ct, gn, and dwt.
Accessories information: Requires two AAA batteries, included.
Ordering information: One-year manufacturers warranty included with purchase.