Popular Science by Celestron SkyMaster 15×17 mm Porro Binoculars
Supplier: Celestron International
71009
470013-340EA 139.89 USD
470013-340
Popular Science by Celestron SkyMaster 15×17 mm Porro Binoculars
Monoculars and Binoculars
Celestron's SkyMaster Series of large aperture binoculars are a phenomenal value for high-performance binoculars ideal for astronomical viewing or for terrestrial use, especially over long distances.
  • 15× magnification porro prism binocular
  • Large 70 mm objective lens offers maximum image brightness in low-light and long-range conditions
  • Ultra-sharp focus across the field of view
  • Multi-coated optics for sharp, clear views
  • Suitable for terrestrial or astronomical viewing
  • Protective rubber covering for ultra-firm grip
  • Large center focus knob for easy focusing
  • Tripod adapter included
  • Long eye relief ideal for eyeglass wearers
Celestron has designed and engineered the SkyMaster models to meet the special demands of extended astronomical or terrestrial viewing sessions.
Carrying case included for travel and safe storage. Limited Lifetime Warranty provides best protection and product support on the market.