Lens and Prism Set
Supplier: American Scientific
3358-20
470013-300CS 574.65 USD
470013-300
Lens and Prism Set
Educational Materials Physics Educational Materials Optics Learning Activities
A Staple For Optic Studies
These sets contain seven different lenses and prisms to demonstrate a wide range of optical principles. The frosted surfaces are also excellent for revealing the path of light as it passes through the lens.