Chemical Spill Tray
Supplier: SCIENTIFIC PLASTICS
T521414201 T523018201 T521714201
470013-236EA 183.05 USD
470013-236 470013-238 470013-240 470200-614
Chemical Spill Tray
Spill Control Materials
Safely Contain Spills!
  • Available in three sizes
  • All trays are 2" deep
One-piece, rotationally-molded polyolefin trays protect cabinets and shelving from acid spills. Corners are rounded for easy cleaning, and walls are 4.8mm (3/16") thick for rigidity. Trays are dishwasher safe and autoclavable.