Pneumatic Trough
Supplier: Humboldt Manufacturing
H-24612
470012-958KT 115.1 USD
470012-958
Pneumatic Trough
Trays/Dishes/Pots Laboratory Trays/Dishes
Stainless Steel with Reinforced Wire Edges.
  • Sloping Sides permit nesting
  • Features overflow tube
Removable sliding shelf is perforated to permit collection of gas samples in inverted gas bottles.