Lab-Aids® Corn Crop Genetics Kit
Supplier: Lab-Aids
603S
470012-426EA 118.05 USD
470012-426
Lab-Aids® Corn Crop Genetics Kit
Educational Materials Biology Educational Materials Genetics Learning Activities
Includes Realistic Corn Ear Cutouts
  • Reusable
  • Uses images of corn ears for consistant results
  • Developed by Sepup
  • Time required: single 50 minute lab
Students build their understanding of selective breeding for desired traits in crops such as corn. They investigate the outcomes of crosses of hybrid and dihybrid corn plants for two unlinked genetic traits: corn kernel color and sweetness. They use Punnett squares to predict the ratio of phenotypes that will be produced by different crosses and analyze the actual ratio of phenotypes of the second generation offspring. A reading describes traditional selective breeding and modern genetic engineering approaches to improving crops to improving crops.
Delivery information: See datasheet for contents information.