How Clean is the Water We Drink and the Air We Breathe? Kit
Supplier: Edvotek
S-30
470012-370EA 75.75 USD
470012-370
How Clean is the Water We Drink and the Air We Breathe? Kit
Educational Materials Biology Educational Materials Microbiology Learning Activities
Make the invisible visible. A safe and simple way to teach pollution and contamination, this kit allows students to detect microscopic microbes in samples of air and water they collect.
  • Collect local air and water samples
  • Observe microorganisms
  • Requires a 400× microscope
Simply incubate the samples overnight, then view the following day with a 400X microscope you provide. Kit includes materials for 10 student groups. Kit requires a microscope with 400× to view the incubated samples.