Thermometer/Hygrometer
Supplier: Taylor Precision Products
497J
470012-294CS 226.15 USD
470012-294
Thermometer/Hygrometer
Hygrometers Analog Hygrometers
Built to Last for Years
With its rustproof parts and shatterproof crystal, this combination thermometer/hygrometer will stand up to years of use. It measures temperature and humidity in any area, indoors or out and the bright graphics are easily visible even from a distance. Relative humidity: 20–90%; temperature range: -50°F to +130°F (-46°C to +54°C). Size: 12" dia.