Ward's® Flame Test and Spectral Analysis Lab Activity
Supplier: Avantor
470012-260KT 103.29 USD
470012-260
Ward's® Flame Test and Spectral Analysis Lab Activity
Educational Classroom Kits and Activities
An Effective Introduction to Spectral Analysis
Astronomers learn important information about the composition of stars by analyzing their spectra. To gain an understanding of this process, students use a spectroscope to observe and describe the spectra of four different chemical salts when heated by a flame. The kit comes with four salts, four nichrome wire test loops, six spectroscopes, a teacher’s guide, and student copymasters. It includes enough materials for 30 or more tests.